Learn More
PISD Monoclonal Antibody (OTI5G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807332S
Encompass_Preferred
 View more versions of this product
Product Code. 17597773
  • € 242.00 / 30µL
Description
PISD Monoclonal antibody specifically detects PISD in Human samples. It is validated for Western Blot
Specifications
Specifications
PISD
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UG56
Mouse
Affinity chromatography
RUO
23761
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI5G2
Unconjugated
PISD
9030221M09Rik; DJ858B16; dJ858B16.2; DKFZp566G2246; phosphatidylserine decarboxylase; Phosphatidylserine decarboxylase alpha chain; Phosphatidylserine decarboxylase beta chain; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; Pisd; PSD; PSDC; PSSC
Human recombinant protein fragment corresponding to amino acids 80-340 of human PISD produced in E.coli.