Learn More
Kinesis Piston seal Thermo yellow TSP 8800 8810 IsochromP-series inert bio
Brand:  Kinesis PS-TSP-602YW
Piston seal Thermo yellow TSP 8800 8810 IsochromP-series inert bio
Product Code. 12914989
  • € 112.00 / Each
Only null left
Estimated Shipment: 03-02-2026