missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pit1 Antibody (PIT1/7262) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20970JF525
This item is not returnable.
View return policy
Description
Pit1 Monoclonal antibody specifically detects Pit1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Pit1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CPHD1, GHF1, GHF-1pituitary-specific positive transcription factor 1, Growth hormone factor 1, PIT-1, PIT1pituitary-specific transcription factor 1, POU class 1 homeobox 1, POU domain class 1, transcription factor 1, POU domain, class 1, transcription factor 1, POU1F1a | |
| Recombinant human full-length Pit1 protein | |
| 0.1 mL | |
| Breast Cancer, Cell Cycle and Replication | |
| 5449 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| PIT1/7262 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction