missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITPNC1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97834JF646
This item is not returnable.
View return policy
Description
PITPNC1 Polyclonal specifically detects PITPNC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PITPNC1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| cytoplasmic phosphatidylinositol transfer protein 1, Mammalian rdgB homolog beta, M-rdgB beta, MrdgBbeta, phosphatidylinositol transfer protein, cytoplasmic 1, RDGBB, RDGBB1, rdgBbeta, RDGB-BETA, retinal degeneration B beta 1, Retinal degeneration B homolog beta | |
| Produced in rabbits immunized with E. coli-derived Human PITPNC1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 26207 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction