missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITX1 Polyclonal Antibody

PITX1 Polyclonal Antibody
Brand: Invitrogen PA5101136
This item is not returnable.
View return policy
Description
PITX1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| PITX1 | |
| Polyclonal | |
| Unconjugated | |
| PITX1 | |
| BFT; CCF; hindlimb expressed homeobox protein backfoot; hindlimb-expressed homeobox protein backfoot; homeobox protein PITX1; Homeobox protein P-OTX; LBNBG; OTX; paired like homeodomain 1; paired-like homeodomain 1; paired-like homeodomain transcription factor 1; pituitary homeo box 1; pituitary homeobox 1; Pituitary OTX-related factor; Pitx1; Potx; P-OTX; PTX1; Unknown (protein for MGC:136976); zgc:136976 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113983, 18740, 5307 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70314, P78337, Q99NA7 | |
| PITX1 | |
| A synthesized peptide derived from human PITX1(Accession P78337), corresponding to amino acid residues R110-Y160. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction