missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PITX2 Polyclonal Antibody

Description
Keep as concentrated solution. Predicted reactivity: Mouse (96%), Rat (95%), Zebrafish (87%), Japanese Medaka (86%), Xenopus laevis (88%), Pig (97%), Chicken (93%), Bovine (97%). Positive Control: PITX2-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pilx2 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. This protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. It plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this protein are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development.
Specifications
Specifications
| Antigen | PITX2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PITX2 |
| Gene Accession No. | Q99697 |
| Gene Alias | 9430085M16Rik; all1-responsive gene 1; ALL1-responsive protein ARP1; Arp1; Brx1; BRX1 homeoprotein; Brx1b; Homeobox protein PITX2; homeodomain transcription factor 2; IDG2; IGDS; IGDS2; IHG2; IRID2; Munc30; orthodenticle-like homeobox 2; Otlx2; paired like homeodomain 2; paired like homeodomain transcription factor 2; paired-like homeodomain 1; paired-like homeodomain 2; paired-like homeodomain transcription factor 1; paired-like homeodomain transcription factor 2; paired-like homeodomain transcription factor 2c; paired-like homeodomain transcription factor Munc 30; Pituitary homeobox 2; PITX1; Pitx2; Pitx-2; Pitx2c; Ptx2; Rgs; Rieg; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; RIEG1; rPtx2; RS; S a 1 glycoprotein; S a1glycoprotein; S alpha 1 glycoprotein; S a 1 glycoprotein; S a1glycoprotein; Solurshin |
| Gene Symbols | PITX2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?