Learn More
Invitrogen™ PITX3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5117477
Product Code. 17269973
  • € 520.00 / 100µg
Description
This antibody has been tested in direct-ELISA.
This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family act as transcription factors. This protein is involved in lens formation during eye development. Mutations of this gene have been associated with anterior segment mesenchymal dysgenesis and congenital cataracts.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PITX3
Polyclonal
Unconjugated
PITX3
ak; aphakia; ASMD; ASOD; CTPP4; CTRCT11; Homeobox protein PITX3; homeobox protein PTX3; MGC12766; paired like homeodomain 3; paired-like homeodomain 3; paired-like homeodomain transcription factor 3; Pituitary homeobox 3; Pituitary homeobox 3 (Homeobox protein PITX3); PITX3; Ptx3
Rabbit
Protein G
RUO
18742, 5309
-20°C or -80°C if preferred
Liquid
Immunohistochemistry (Paraffin), Western Blot
3 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
O35160, O75364
PITX3
Recombinant Human Pituitary homeobox 3 protein (10-200aa).
100 μg
Primary
Human, Mouse