missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITX3 Recombinant Rabbit Monoclonal Antibody (070)

PITX3 Recombinant Rabbit Monoclonal Antibody (70)
Brand: Invitrogen MA530757
This item is not returnable.
View return policy
Description
Specifications
| PITX3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O75364 | |
| PITX3 | |
| Recombinant Human PTX3/Pentraxin 3/TSG-14 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 070 | |
| Unconjugated | |
| PITX3 | |
| ak; aphakia; ASMD; ASOD; CTPP4; CTRCT11; Homeobox protein PITX3; homeobox protein PTX3; MGC12766; paired like homeodomain 3; paired-like homeodomain 3; paired-like homeodomain transcription factor 3; Pituitary homeobox 3; Pituitary homeobox 3 (Homeobox protein PITX3); PITX3; Ptx3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5309 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction