missing translation for 'onlineSavingsMsg'
Learn More

Pivampicillin Hydrochloride, LoGiCal

Product Code. 30463606
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463606 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463606 Supplier LoGiCal Supplier No. LGCFOR3237.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pivampicillin Hydrochloride
CAS 26309-95-5
Analyte or Component Names Pivampicillin Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C22 H29 N3 O6 S . Cl H
Certifications/Compliance ISO 9001
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, hydrochloride (1:1), (2S,5R,6R), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-(2-amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-, hydroxymethyl ester pivalate (ester), monohydrochloride, D- (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[(aminophenylacetyl)amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, monohydrochloride, [2S-[2α,5α,6β(S*)]]-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(2R)-aminophenylacetyl]amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, monohydrochloride, (2S,5R,6R)- (9CI), Methanediol, 6-(2-amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate pivalate (ester) monohydrochloride (8CI), Pivalic acid, hydroxymethyl ester 6-(2-amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate monohydrochloride (8CI), Alphacilina, Alphacillin, Ampicillin pivaloxyoxymethyl ester hydrochloride, Ampicillin pivaloyloxymethyl ester hydrochloride, Berocillin, Centurina, Diancina, Inacilin, Maxifen, Pivaloyloxymethyl D-(-)-α-aminophenylacetamidopenicillanate hydrochloride, Pivampicillin hydrochloride, Pivatil, Pondocil, Pondocillin, Pondocillina, Sanguicillin
InChI Formula InChI=1S/C22H29N3O6S.ClH/c1-21(2,3)20(29)31-11-30-19(28)15-22(4,5)32-18-14(17(27)25(15)18)24-16(26)13(23)12-9-7-6-8-10-12;/h6-10,13-15,18H,11,23H2,1-5H3,(H,24,26);1H/t13-,14-,15+,18-;/m1./s1
Solution Type Neat
SMILES Cl.CC(C)(C)C(=O)OCOC(=O)[C@@H]1N2[C@H](SC1(C)C)[C@H](NC(=O)[C@H](N)c3ccccc3)C2=O
IUPAC Name 2,2-dimethylpropanoyloxymethyl (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 500.01
Quantity 5 mg
Formula Weight 499.1544 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.