Learn More
Pivmecillinam Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Pivmecillinam Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Pivmecillinam hydrochloride |
| CAS | 32887-03-9 |
| Analyte or Component Names | Pivmecillinam hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C21 H33 N3 O5 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, hydrochloride (1:1), (2S,5R,6R), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, monohydrochloride, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, hydroxymethyl ester pivalate (ester), monohydrochloride, (+)- (8CI), Melysin, Pivmecillinam hydrochloride, Selexid, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, monohydrochloride, [2S-(2alpha,5alpha,6beta)]- |
| InChI Formula | InChI=1S/C21H33N3O5S.ClH/c1-20(2,3)19(27)29-13-28-18(26)15-21(4,5)30-17-14(16(25)24(15)17)22-12-23-10-8-6-7-9-11-23;/h12,14-15,17H,6-11,13H2,1-5H3;1H/b22-12+;/t14-,15+,17-;/m1./s1 |
| SMILES | Cl.CC(C)(C)C(=O)OCOC(=O)[C@@H]1N2[C@H](SC1(C)C)[C@H](\N=C\N3CCCCCC3)C2=O |
| IUPAC Name | 2,2-dimethylpropanoyloxymethyl (2S,5R,6R)-6-[(E)-azepan-1-ylmethylideneamino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrochloride |
| Show More |