Learn More
Invitrogen™ PIWIL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140240
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DLAVHTRLTP EQRQREVGRL IDYIHKNDNV QRELRDWGLS FDSNLLSFSG RILQTEKIHQ GGKTFDYNPQ FADWSKETRG APLISVKPL Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.
Hiwi is a member of the Piwi /Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34- hematopoietic cells, suggesting a role for this gene in the maintenance of hematopoietic stem cells.
Specifications
| PIWIL1 | |
| Polyclonal | |
| Unconjugated | |
| PIWIL1 | |
| CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9271 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96J94 | |
| PIWIL1 | |
| Recombinant protein corresponding to Human PIWIL1. Recombinant protein control fragment (Product #RP-92293). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.