Learn More
Invitrogen™ PIWIL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519457
Description
PA5-19457 targets PIWIL1 in WB applications and shows reactivity with mouse and Rat samples. The PA5-19457 immunogen is synthetic peptide derived from within residues 800 - 900 of Human PIWIL1. PA5-19457 detects PIWIL1 which has a predicted molecular weight of approximately 99 kDa.
Hiwi is a member of the Piwi /Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34- hematopoietic cells, suggesting a role for this gene in the maintenance of hematopoietic stem cells.
Specifications
| PIWIL1 | |
| Polyclonal | |
| Unconjugated | |
| PIWIL1 | |
| CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 363912, 57749, 9271 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| Q96J94, Q9JMB7 | |
| PIWIL1 | |
| Synthetic peptide derived from within residues 800 - 900 of Human PIWIL1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.