missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIWIL2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41143AF647
This item is not returnable.
View return policy
Description
PIWIL2 Polyclonal antibody specifically detects PIWIL2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PIWIL2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 647 | |
| Cancer/testis antigen 80, CT80mili, FLJ10351, HILIMiwi like, MGC133049, Mili, PIWIL1L, piwil2-like protein, piwi-like 2 (Drosophila), piwi-like protein 2 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human PIWI-L2. The immunogen is located within amino acids 410 - 460 of PIWI-L2. Amino Acid Squence: TRYNNRTYRIDDVDWNK | |
| 0.1 mL | |
| Stem Cells | |
| 55124 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction