missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pixantrone Dimaleate, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Pixantrone Dimaleate |
| CAS | 144675-97-8 |
| Molecular Formula | C17 H19 N5 O2 . 2 C4 H4 O4 |
| Quantity | 10 mg |
| Synonym | Benz[g]isoquinoline-5,10-dione','6,9-bis[(2-aminoethyl)amino]-','(2Z)-2-butenedioate (1:2), Benz[g]isoquinoline-5,10-dione','6,9-bis[(2-aminoethyl)amino]-','(Z)-2-butenedioate (1:2), BBR 2778, Pixantrone dimaleate, Pixantrone maleate |
| InChI Formula | InChI=1S/C17H19N5O2.2C4H4O4/c18-4-7-21-12-1-2-13(22-8-5-19)15-14(12)16(23)10-3-6-20-9-11(10)17(15)24;2*5-3(6)1-2-4(7)8/h1-3,6,9,21-22H,4-5,7-8,18-19H2;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1- |
| SMILES | NCCNc1ccc(NCCN)c2C(=O)c3cnccc3C(=O)c12.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O |
| IUPAC Name | 6,9-bis(2-aminoethylamino)benzo[g]isoquinoline-5,10-dione;(Z)-but-2-enedioic acid |
| Molecular Weight (g/mol) | 557.51 |
| Formula Weight | 557.1758 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction