missing translation for 'onlineSavingsMsg'
Learn More

Pixantrone Dimaleate, TRC

Product Code. 30426740
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30426740

Brand: Toronto Research Chemicals TRCP552500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pixantrone Dimaleate
CAS 144675-97-8
Molecular Formula C17 H19 N5 O2 . 2 C4 H4 O4
Quantity 100 mg
Synonym Benz[g]isoquinoline-5,10-dione','6,9-bis[(2-aminoethyl)amino]-','(2Z)-2-butenedioate (1:2), Benz[g]isoquinoline-5,10-dione','6,9-bis[(2-aminoethyl)amino]-','(Z)-2-butenedioate (1:2), BBR 2778, Pixantrone dimaleate, Pixantrone maleate
InChI Formula InChI=1S/C17H19N5O2.2C4H4O4/c18-4-7-21-12-1-2-13(22-8-5-19)15-14(12)16(23)10-3-6-20-9-11(10)17(15)24;2*5-3(6)1-2-4(7)8/h1-3,6,9,21-22H,4-5,7-8,18-19H2;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
SMILES NCCNc1ccc(NCCN)c2C(=O)c3cnccc3C(=O)c12.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O
IUPAC Name 6,9-bis(2-aminoethylamino)benzo[g]isoquinoline-5,10-dione;(Z)-but-2-enedioic acid
Molecular Weight (g/mol) 557.51
Formula Weight 557.1758
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.