Learn More
Pixantrone, MedChemExpress
Product Code. 16481977
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
50mg
5mg
Product Code. 16481977
missing translation for 'mfr': Medchem Express HY13727A
MedChemExpress Pixantrone (BBR 2778) dimaleate is a topoisomerase II inhibitor and DNA intercalator, with anti-tumor activity.
Pixantrone (BBR 2778) dimaleate is a topoisomerase II inhibitor and DNA intercalator, with anti-tumor activity.
Specifications
Chemical Name or Material Pixantrone
CAS 144675-97-8
Color Blue
Purity Grade Notes Research
Molecular Formula C25H27N5O10
Quantity 50 mg
Synonym BBR 2778
Solubility Information DMSO : 33.33 mg/mL (59.78 mM; Need ultrasonic) ∣H2O : 5 mg/mL (8.97 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(O)/C=C\C(O)=O.O=C(O)/C=C\C(O)=O.O=C(C1=C(NCCN)C=CC(NCCN)=C12)C3=C(C=NC=C3)C2=O
Molecular Weight (g/mol) 557.51
Formula Weight 557.51
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only