missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PJA Polyclonal Antibody

Product Code. 15709380 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15709380 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15709380 Supplier Invitrogen™ Supplier No. PA551419

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LPARPSRAPV SICGGGENTS KSAEEPVVRP KIRNLASPNC VKPKIFFDTD DDDDMPHSTS RWRDTANDNE GHSDGLARRG RGESSSGYPE PKYPEDKREA RSDQVKP Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 60%.

Has E2-dependent E3 ubiquitin-protein ligase activity. Ubiquitinates MAGED1 antigen leading to its subsequent degradation by proteasome. May be involved in protein sorting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PJA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PJA1
Gene Accession No. Q8NG27
Gene Alias E3 ubiquitin-protein ligase Praja-1; Pja1; praja 1; praja ring finger 1, E3 ubiquitin protein ligase; praja ring finger ubiquitin ligase 1; Praja1; praja-1; praja1, RING-H2 motif containing; RING finger protein 70; RING-type E3 ubiquitin transferase Praja-1; RNF70
Gene Symbols PJA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PJA. Recombinant protein control fragment (Product #RP-91978).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64219
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.