missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKA beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117489
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
The catalytic subunit C-beta of PKA (PRKACB) is a member of the Ser/Thr protein kinase family (the PKA catalytic subunit consist of three gene products: C-alpha, C-beta, and C-gamma) and has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans.
Specifications
| PKA beta | |
| Polyclonal | |
| Unconjugated | |
| PRKACB | |
| cAMP-dependent protein kinase C beta; cAMP-dependent protein kinase catalytic beta subunit isoform 4ab; cAMP-dependent protein kinase catalytic subunit beta; PKA C-beta; Pkacb; PRKACB; protein kinase A catalytic subunit beta; protein kinase cAMP-activated catalytic subunit beta; protein kinase, cAMP dependent, catalytic, beta; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, beta | |
| Rabbit | |
| Protein G | |
| RUO | |
| 18749, 293508, 5567 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P22694, P68181, P68182 | |
| PRKACB | |
| Recombinant Human cAMP-dependent protein kinase catalytic subunit beta protein (2-53aa). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction