missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKA C-gamma Antibody (2E4), Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | PKA C-gamma |
|---|---|
| Clone | 20000 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18109787
|
Novus Biologicals
NBP2-37487 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688705
|
Novus Biologicals
NBP2-37487-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKA C-gamma Monoclonal specifically detects PKA C-gamma in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PKA C-gamma | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| cAMP-dependent protein kinase catalytic subunit gamma, EC 2.7.11, EC 2.7.11.11, KAPG, PKA C-gamma, PKACg, protein kinase, cAMP-dependent, catalytic, gamma, serine(threonine) protein kinase | |
| PRKACG | |
| IgG1 | |
| Unpurified |
| 20000 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Protein Kinase, Wnt Signaling Pathway | |
| Ascites with 0.03% Sodium Azide | |
| 5568 | |
| Purified recombinant fragment of human PRKACG expressed in E. Coli. | |
| Primary | |
| 40.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title