missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKA R2, p Ser99 Antibody (SR1905), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21511-50ul
This item is not returnable.
View return policy
Description
PKA R2 Monoclonal antibody specifically detects PKA R2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| PKA R2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1905 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| protein kinase, cAMP-dependent, regulatory, type II, alpha, RII-alpha subunit | |
| A synthesized peptide derived from human phospho-PKA R2 [Ser99] (Uniprot #: P13861) | |
| 50 μg | |
| Phospho Specific, Signal Transduction | |
| 5576 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction