missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKA RI beta Antibody (OTI2A3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73430H
This item is not returnable.
View return policy
Description
PKA RI beta Monoclonal specifically detects PKA RI beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PKA RI beta | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2A3 | |
| Western Blot, Immunohistochemistry | |
| PRKAR1B | |
| Full length recombinant protein of human PKA regulatory subunit I beta (NP_002726) produced in HEK293T cells. | |
| 0.1 mL | |
| 5575.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction