missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human PTH 15N Stable Isotope Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
Description
Test in Process.
Specifications
Specifications
| Accession Number | P01270, P01270, P01270 |
| For Use With (Application) | PAGE |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 5741 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 8.9 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of PTH as determined by LAL method. |
| Gene Alias | Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 |
| Gene Symbol | PTH |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction