missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC Alpha, Beta, Gamma Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 190.00 - € 507.00
Specifications
| Antigen | PKC Alpha, Beta, Gamma |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232043
|
Novus Biologicals
NBP3-33469-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228079
|
Novus Biologicals
NBP3-33469-100ul |
100 μL |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKC Alpha, Beta, Gamma Monoclonal antibody specifically detects PKC Alpha, Beta, Gamma in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| PKC Alpha, Beta, Gamma | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| PKC (a,b,y), PKC a, b, g, PKC Alpha, PKC Alpha Beta Gamma, PKC Beta, PKC Gamma | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| A synthetic peptide corresponding to a sequence within amino acids 300-500 of human pan PKC (NP_002728.2/NP_002729.2/NP_002730.1).,, Sequence:, ELRQKFERAKISQGTKVPEEKTTNTVSKFDNNGNRDRMKLTDFNFLMVLGKGSFGKVMLSERKGTDELYAVKILKKDVVIQDDDVECTMVEKRVLALPGKPPFLTQLHSCFQTMDRLYFVMEYVNGGDLMYHIQQVGRFKEPHAVFYAAEIAIGLFFLQSKGIIYRDLKLDNVMLDSEGHIKIADFGMCKENIWDGVTTKT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title