Learn More
Novus Biologicals™ PKC beta 2 Antibody (SR2824), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27716-50ul
 View more versions of this product
Product Code. 30041693
  • € 316.00 / 50µL
Description
PKC beta 2 Monoclonal antibody specifically detects PKC beta 2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PKC beta 2
Monoclonal
Unconjugated
PBS (pH 7.4), 150mM NaCl, 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
SR2824
Western Blot 1:500-1:2000, Flow Cytometry, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
PKCB;PKCbeta;PKC-beta;PKCI(2);PRKCB;PRKCB1;PRKCB2;Protein kinase C beta type
A synthesized peptide derived from human PKC beta 2 (Uniprot #: P05771)
50 μL
Cancer, mTOR Pathway, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
5579
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.