missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKC delta Monoclonal Antibody (K06341_6F1), Invitrogen™
Mouse Monoclonal Antibody
€ 538.00 - € 900.00
Specifications
| Antigen | PKC delta |
|---|---|
| Clone | K06341_6F1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30210947
|
Invitrogen™
MA556327 |
200 μL |
€ 900.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30199021
|
Invitrogen™
MA555947 |
50 μL |
€ 538.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: KCREKVANLC GINQKLLAEA LNQVTQRASR RSDSASSEPV GIYQGFEKKT GVAGEDMQDN SGTYGKIWEG
PKC delta Monoclonal antibody specifically detects PKC delta in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| PKC delta | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1-May; AI385711; ALPS3; CVID9; D14Ertd420e; kinase nPKC-delta; KPCD; MAY1; nPKC-delta; PKC[d]; Pkcd; PKCdelta; Prkcd; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C[d]; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD | |
| Recombinant human PRKCD (immunogen range is 271-340). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PRKCD |
| K06341_6F1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q05655 | |
| 5580 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title