Learn More
PKC eta Antibody (SR1663), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21825-100ul
This item is not returnable.
View return policy
Description
PKC eta Monoclonal antibody specifically detects PKC eta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PKC eta | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1663 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| EC 2.7.11, EC 2.7.11.13, nPKC-eta, PKCLMGC26269, PKC-LMGC5363, PRKCLprotein kinase C eta type, protein kinase C, eta | |
| A synthesized peptide derived from human PKC eta (Uniprot #: P24723) | |
| 100 μg | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 5583 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |