missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKC eta Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5119589

Product Code. 17720994

  • € 488.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PKC eta
Polyclonal
Unconjugated
PRKCH
LOW QUALITY PROTEIN: protein kinase C eta type; MGC26269; MGC5363; nPKC-eta; PKC; Pkch; PKCL; PKC-L; PRKCH; PRKCL; protein kinase C eta; protein kinase C eta type; protein kinase C eta type pseudogene; protein kinase C theta; protein kinase C, eta; protein kinase C-eta; Unknown (protein for MGC:140784)
Rabbit
Antigen affinity chromatography
RUO
18755
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1.07 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
P23298
PRKCH
Recombinant protein encompassing a sequence within the C-terminus region of mouse PKC eta. The exact sequence is proprietary.
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.