missing translation for 'onlineSavingsMsg'
Learn More

PKC mu [p Ser916] Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 190.00 - € 724.00

Specifications

Antigen PKC mu
Dilution Western Blot 1:500 - 1:2000, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229820
View Documents
Novus Biologicals
NBP3-38645-100ul
100 μL
€ 724.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30231455
View Documents
Novus Biologicals
NBP3-38645-20ul
20 μL
€ 190.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PKC mu Polyclonal antibody specifically detects PKC mu in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications

Specifications

PKC mu
ELISA, Western Blot
Unconjugated
Rabbit
Golgi Apparatus Markers, Phospho Specific, Signal Transduction
PBS (pH 7.3), 50% glycerol
5587
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA
Polyclonal
Purified
RUO
Human, Mouse
EC 2.7.11, nPKC-D1, nPKC-mu, PKCM, PKC-MU, PKD1, PKDEC 2.7.11.13, PRKCMPKC-mu, Protein kinase C mu type, protein kinase C, mu, Protein kinase D, protein kinase D1, serine/threonine-protein kinase D1
A synthetic phosphorylated peptide around S912 of human PKC mu (NP_002733.2).,, Sequence:, RVSIL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.