Learn More
Eurofins DiscoverX PKC ζ Protein, Active
Description
KPCZ (Protein kinase C, zeta type, nPKC-zeta, EC 2.7.1.37) is one of the major protein kinase C (PKC) isoforms of a family of serine/threonine protein kinases. Breakdown of phosphatidylinositol leads to formation of diacylglycerol and inositol triphophate, the former acting synergistically with Ca2+ to activate PKC, resulting in translocation of the enzyme from cytosol to the membrane. Phosphorylation of proteins by PKC has been implicated as a pivotal regulatory element in signal transduction, cellular regulation, and with a high affinity to phorbol ester tumor promoters.
GenBank BC014270
Specifications
Specifications
| Formulation | mg/mL of enzyme in 20 mM Tris/HCl pH 7.5, 0.02% Triton X-100, 5% glycerol, 1 mM EDTA, 1 mM EGTA, 0.1% 2-mercaptoethanol, 1 mM PMSF, 10 mM benzamidine. Frozen solution. |
| Molecular Weight (g/mol) | 68.9 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥73% |
| Protein | PKCζ (zeta), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.