Learn More
Invitrogen™ PKC theta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596442
Description
Immunogen sequence: MSPFLRIGLS NFDCGSCQSC QGEAVNPYCA VLVKEYVESE NGQMYIQKKP TMYPPWDSTF DAHINKGRVM QIIVKGKNVD LISETTVELY SLAERCRKNN GKTEIWLELK PQGRMLMNAR YFLEMSDTKD MNEFETEGFF ALHQRRGAIK QAKVHHVKCH EFTATFFPQP TFCSVCHEFV WGLNKQGYQC RQCNAAIHKK CIDKVIAKCT GSAINSRETM FHKERFKIDM; Positive Samples: HepG2, Rat testis; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein.
Protein Kinase C (PKC) isoforms are serine/threonine kinases involved in signal transduction pathways that govern a wide range of physiological processes including differentiation, proliferation, gene expression, brain function, membrane transport and the organization of cytoskeletal and extracellular matrix proteins. Increasing evidence from studies using in vitro and in vivo systems points to PKC as a key regulator of critical cell cycle transitions, including cell cycle entry and exit and the G1 and G2 checkpoints. PKC-mediated control of these transitions can be negative or positive, depending on the timing of PKC activation during the cell cycle and on the specific PKC isozymes involve. There have been at least 12 different PKC isoforms identified in humans to date, including alpha, beta I, beta II, gamma, delta, epsilon, zeta, eta, theta, iota, lambda, and mu. The different PKC isoforms vary in structure, subcellular localization, tissue distribution and response to extracellular signals. PKC beta II has been shown to be expressed, in part, in cardiac muscle, intestinal epithelium, kidney and pancreas. PKC beta II has been shown to be involved in cell cycle progression and cancer proliferation.
Specifications
| PKC theta | |
| Polyclonal | |
| Unconjugated | |
| PRKCQ | |
| A130035A12Rik; AW494342; EC 2.7.11.13; kinase nPKC-theta; KPCT; MGC126514; MGC141919; nPKC-theta; PKC theta; PKC-0; Pkcq; PKCtheta; PKC-theta; PRKCQ; PRKCT; protein kinase C theta; protein kinase C theta type; protein kinase C, theta; RP11-563J2.1; si:ch211-239o15.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18761, 5588, 85420 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02111, Q04759, Q9WTQ0 | |
| PRKCQ | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human PRKCQ (NP_0062481). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.