missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKD1L3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17989082
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17989082 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17989082 Supplier Invitrogen™ Supplier No. PA5144243

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PKD1L3 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen PKD1L3
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene PKD1L3
Gene Accession No. Q7Z443
Gene Alias PC1-like 3 protein; PK1L3; PKD1L3; polycystic kidney disease 1-like 3; polycystic kidney disease protein 1-like 3; polycystin 1 like 3, transient receptor potential channel interacting; polycystin-1L3
Gene Symbols PKD1L3
Host Species Rabbit
Immunogen Recombinant Human Polycystic kidney disease protein 1-like 3 protein (22-194AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342372
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.