Learn More
Invitrogen™ PKD2 Monoclonal Antibody (K49022_2E5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556070
Description
Sequence of this protein is as follows: SDLIRKGYHK ALVKLKLKKN TVDDISESLR QGGGKLNFDE LRQDLKGKGH TDAEIEAIFT KYDQDGDQEL TEHEHQQMRD DLEKEREDLD LDHSSLPRPM SSRSFPRSLD DSEEDDDEDS GHSSRRRGSI SSGVSYEEFQ VLVRRVDRME HSIGSIVSKI DAVIVKLEIM ERAKLKRREV LGRLLDGVAE DERLGRDSEI HREQMERLVR EELERWESDD AASQISHGLG TPVGLNGQPR PRSSRPSSSQ
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal dominant polycystic kidney disease.
Specifications
| PKD2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O35245, Q13563 | |
| PKD2 | |
| Recombinant human PKD2 (immunogen range is 701-950). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K49022_2E5 | |
| Unconjugated | |
| PKD2 | |
| A MGC138466; APKD2; Autosomal dominant polycystic kidney disease type II protein; C030034P18Rik; MGC138468; PC2; Pc-2; PKD2; PKD4; polycystic kidney disease 2; polycystic kidney disease 2 (autosomal dominant); polycystic kidney disease 2 homolog; Polycystic kidney disease 2 protein; Polycystic kidney disease 2 protein homolog; polycystin 2; polycystin 2, transient receptor potential cation channel; polycystin-2; Polycystwin; R48321; RGD1559992; transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2; TRPP1; TRPP2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 18764, 5311 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.