missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX PKG1α Protein, Active

Product Code. 17360564
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17360564

Brand: Eurofins DiscoverX 14688

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal 6His-tagged, recombinant, full length PKG1α, for use in Kinase Assays.

In smooth muscle cells, one of the major receptors for cGMP is the serine/threonine protein kinase, cGMP-dependent protein kinase 1, beta isozyme (PRKG1B). PRKG1 has been shown to catalyze the phosphorylation of a number of physiologically relevant proteins whose function it is to regulate the contractile activity of the smooth muscle cell. Two main types of PRKG are present in eukaryotic cells. Type I is widely distributed and is isolated from soluble extracts of tissues, such as cardiac and smooth muscle, as well as lymphoid cells. Type II is a particulate form of the enzyme and has a limited tissue distribution.

GenBank D45864

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 80 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥64%
Protein PKG1α, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.