missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKHD1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals BC110-60480AF488
This item is not returnable.
View return policy
Description
PKHD1 Polyclonal specifically detects PKHD1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| PKHD1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence, Peptide ELISA | |
| P08F94 | |
| PKHD1 | |
| A synthetic peptide derived from the human PKHD1 protein (within residues 3800-3900). [UniProt# Q8TCZ9] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Immunocytochemistry, Immunofluorescence, ELISA | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| polycystic kidney and hepatic disease 1 (autosomal recessive) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5314 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction