missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKIA Mouse anti-Human, Clone: OTI1G5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810311
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the cAMP-dependent protein kinase (PKA) inhibitor family. This protein was demonstrated to interact with and inhibit the activities of both C alpha and C beta catalytic subunits of the PKA. Alternatively spliced transcript variants encoding the same protein have been reported.Specifications
| PKIA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P61925 | |
| PKIA | |
| Full length human recombit protein of human PKIA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1G5 | |
| Unconjugated | |
| PKIA | |
| PRKACN1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5569 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction