missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKIB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570346
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 90%; Dog: 90%; Goat: 85%; Guinea Pig: 92%; Horse: 90%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a member of the cAMP-dependent protein kinase inhibitor family. The encoded protein may play a role in the protein kinase A (PKA) pathway by interacting with the catalytic subunit of PKA, and overexpression of this gene may play a role in prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Jul 2012].
Specifications
| PKIB | |
| Polyclonal | |
| Unconjugated | |
| PKIB | |
| cAMP-dependent protein kinase (catalytic subunit binding) inhibtor 2; cAMP-dependent protein kinase inhibitor 2; cAMP-dependent protein kinase inhibitor beta; cAMP-dependent protein kinase inhibitor bet-like protein; cAMP-dependent protein kinase inhibitor, testis isoform; im:7054531; Pkib; Pkibeta; PKI-beta; PKINH; PRKACN2; protein kinase (cAMP dependent, catalytic) inhibitor beta; protein kinase (cAMP-dependent, catalytic) inhibitor beta; protein kinase inhibitor beta, (cAMP-dependent, catalytic) inhibitor beta; protein kinase inhibitor beta, cAMP dependent, catalytic; protein kinase inhibitor beta, cAMP dependent, testis specific; protein kinase inhibitor, beta; protein kinase, cAMP-dependent, catalytic, inhibitor 2; RATPKINH; si:dkey-150h13.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24678 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P27775 | |
| PKIB | |
| synthetic peptide corresponding to a region of rat PKIB (aa 2-51). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction