Learn More
PKLR Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97071F
Product Code. 18086166
  • € 415.00 / 0.10mL
Description
PKLR Polyclonal specifically detects PKLR in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PKLR
Polyclonal
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
EC 2.7.1.40, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase
Produced in rabbits immunized with purified, recombinant PKLR (NP_000289.1; Leu47-Ser574).
0.1 mL
Chaperone Mediated Autophagy (CMA), Lipid and Metabolism, Signal Transduction
5313
Store at 4°C in the dark.
IgG
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS
Rabbit
Affinity purified