missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKLR Monoclonal Antibody (OTI1H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
POA
Specifications
| Antigen | PKLR |
|---|---|
| Clone | OTI1H9 |
| Concentration | 1 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17504181
|
Origene Technologies
TA501414S |
30 μL |
POA
30µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17584171
|
Origene Technologies
TA501414 |
100 μL |
POA
100µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PKLR Monoclonal antibody specifically detects PKLR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| PKLR | |
| 1 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| 5313 | |
| Primary | |
| PKLR |
| OTI1H9 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P30613 | |
| IgG2a | |
| -20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title