Learn More
PKLR Mouse anti-Human, Clone: OTI1F3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501412
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a pyruvate kinase that catalyzes the transphosphorylation of phohsphoenolpyruvate into pyruvate and ATP, which is the rate-limiting step of glycolysis. Defects in this enzyme, due to gene mutations or genetic variations, are the common cause of chronic hereditary nonspherocytic hemolytic anemia (CNSHA or HNSHA). Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| PKLR | |
| Monoclonal | |
| Unconjugated | |
| PKLR | |
| PK1, PKL, PKR, PKRL, RPK | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5313 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1F3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P30613 | |
| PKLR | |
| Full length human recombit protein of human PKLR produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.