missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKM1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14833V
This item is not returnable.
View return policy
Description
PKM1 Polyclonal specifically detects PKM1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications
| PKM1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting | |
| PK1, Pyruvate kinase isozymes M1, Pyruvate kinase M1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| This antibody is specific for the M1 isoform of PKM. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PKM | |
| A synthetic peptide made to a C-terminal portion of the human PKM1 protein (between residues 350-450) [UniProt P14618-2]. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Bovine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction