Learn More
PKM2 Mouse anti-Human, Clone: OTI12D5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA190266
Description
Synthetic peptide sequence is LRRLAPITSDPTEATAVGAV
PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.Specifications
| PKM2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P14618 | |
| PKM | |
| Synthetic peptide corresponding to the region encoded by PKM gene M2 isoform. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI12D5 | |
| Unconjugated | |
| PKM | |
| PKM2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5315 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.