missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKMYT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517665
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
PKMYT1 serine/threonine kinase is a member of a family of neural specific, zinc finger-containing DNA-binding proteins which binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. MYT1 of the oligodendrocyte lineage, along with a closely related CCHC zinc finger, is expressed in developing neurons in the mammalian central nervous system. The recombinant PKMYT1 fragments containing either the upstream 2 zinc fingers or the downstream 4 zinc fingers bound the same cis regulatory element in the PLP1 promoter in vitro and DNA binding required Zn2+, but not other divalent cations.
Specifications
| PKMYT1 | |
| Polyclonal | |
| Unconjugated | |
| Pkmyt1 | |
| 6230424P17; AW209059; Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; membrane-associated tyrosine-and threonine-specific cdc2-inhibitory kinase; Myt1; Myt1 kinase; Pkmyt1; pkmyt1.S; PPP1R126; protein kinase, membrane associated tyrosine/threonine 1; protein kinase, membrane associated tyrosine/threonine 1 S homeolog; protein phosphatase 1, regulatory subunit 126; RGD1305434; XELAEV_18047881mg | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 268930, 287101, 379626, 9088 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 30 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q91618, Q99640, Q9ESG9 | |
| Pkmyt1, pkmyt1.S | |
| Synthetic peptide corresponding to residues near the middle of mouse and human Myt1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Frog | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction