missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKN3 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30102AF488
This item is not returnable.
View return policy
Description
PKN3 Polyclonal specifically detects PKN3 in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.
Specifications
| PKN3 | |
| Polyclonal | |
| Western Blot, Knockout Validated | |
| EC 2.7.11, EC 2.7.11.13, PKNbeta, protein kinase N3, Protein kinase PKN-beta, Protein-kinase C-related kinase 3, RP11-545E17.1, serine/threonine-protein kinase N3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoassay | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PKN3 | |
| maps to a region between residue 225 and 275 of human protein kinase N3 using the numbering given in entry NP_037487.2 | |
| 100 μL | |
| Protein Kinase | |
| 29941 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction