missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKN3 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98608UV
This item is not returnable.
View return policy
Description
PKN3 Polyclonal antibody specifically detects PKN3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PKN3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| EC 2.7.11, EC 2.7.11.13, PKNbeta, protein kinase N3, Protein kinase PKN-beta, Protein-kinase C-related kinase 3, RP11-545E17.1, serine/threonine-protein kinase N3 | |
| Produced in rabbits immunized with E. coli-derived Human PKN3 fragment. | |
| 0.1 mL | |
| Protein Kinase | |
| 29941 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction