missing translation for 'onlineSavingsMsg'
Learn More

PKNOX1 Antibody, Novus Biologicals™

Product Code. 30229626 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30229626 20 μL 20µL
30231562 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30229626 Supplier Novus Biologicals Supplier No. NBP33552520ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PKNOX1 Polyclonal antibody specifically detects PKNOX1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKNOX1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:2000, ELISA
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias Homeobox protein PREP-1, PBX/kn10Pbx regulating protein-1, pkonx1c
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 322-436 of human PKNOX1 (NP_004562.2).,, Sequence:, LDSSCSETPKTKKKTAQNRPVQRFWPDSIASGVAQPPPSELTMSEGAVVTITTPVNMNVDSLQSLSSDGATLAVQQVMMAGQSEDESVDSTEEDAGALAPAHISGLVLENSDSLQ
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5316
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.