missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKNOX1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566065

Product Code. 15908794

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ATQTLSIDSYQ DGQQMQVVTE LKTEQDPNCS EPDAEGVSPP Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

PKNOX1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PKNOX1
Polyclonal
Unconjugated
Pknox1
D17Wsu76e; homeobox protein PKNOX1; Homeobox protein PREP-1; human homeobox-containing protein; Pbx regulating protein-1; Pbx/knotted 1 homeobox; PBX/knotted 1 homeobox 1; PBX/knotted homeobox 1; PKNOX1; pkonx1c; PREP1
Rabbit
Antigen affinity chromatography
RUO
5316
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P55347
Pknox1
Recombinant Human PKNOX1. Recombinant protein control fragment (Product #RP-106758).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.