missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKP4 Polyclonal Antibody

PKP4 Polyclonal Antibody
Brand: Invitrogen PA566855
This item is not returnable.
View return policy
Description
PKP4 Polyclonal Antibody for ICC/IF
Specifications
| PKP4 | |
| Polyclonal | |
| Unconjugated | |
| PKP4 | |
| 5031422I09Rik; 9430019K17Rik; Armadillo-related protein; Armrp; catenin 4; p0071; PKP4; plakophilin 4; plakophilin-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8502 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99569 | |
| PKP4 | |
| Recombinant Human PKP4. Recombinant protein control fragment (Product #RP-107509). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction