missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLA2 Monoclonal Antibody (630823)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA524282

Product Code. 15553972

  • € 475.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

In direct ELISAs, no cross-reactivity with recombinant human (rh) PLA2G2A, rhPLA2G7, recombinant mouse (rm) PLA2G2A, or rmPLA2R1 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Phospholipase A2 catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLA2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P04054
PLA2G1B
Mouse myeloma cell line NS0-derived recombinant human PLA2G1B Met1-Ser148.
100 μg
Primary
Human
Antibody
IgG2a
Immunoprecipitation, Western Blot
630823
Unconjugated
PLA2G1B
Group IB phospholipase A2; group IB secretory phospholipase A(2); MGC119834; MGC119835; phosphatidylcholine 2-acylhydrolase 1B; phospholipase; phospholipase A2; phospholipase A2 group IB; phospholipase A2, group 1B; phospholipase A2, group IB (pancreas); phospholipase A2, group IB, pancreas; phospholipase A2, major isoenzyme; PLA2; PLA2A; Pla2g1b; PLA2-Ib; PPLA2; sPLA(2)-IB; sPLA2IB; unnamed protein product
Mouse
Protein A/G
RUO
5319
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.