missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584468
This item is not returnable.
View return policy
Description
Immunogen sequence: ADSGISPRAV WQFRKMIKCV IPGSDPFLEY NNYGCYCGL Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 79%.
Phospholipase A2 catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets.
Specifications
| PLA2 | |
| Polyclonal | |
| Unconjugated | |
| PLA2G1B | |
| Group IB phospholipase A2; group IB secretory phospholipase A(2); MGC119834; MGC119835; phosphatidylcholine 2-acylhydrolase 1B; phospholipase; phospholipase A2; phospholipase A2 group IB; phospholipase A2, group 1B; phospholipase A2, group IB (pancreas); phospholipase A2, group IB, pancreas; phospholipase A2, major isoenzyme; PLA2; PLA2A; Pla2g1b; PLA2-Ib; PPLA2; sPLA(2)-IB; sPLA2IB; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P04054 | |
| PLA2G1B | |
| Recombinant protein corresponding to Human PLA2G1B. Recombinant protein control fragment (Product #RP-99897). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction