missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2G12A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569189
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s.
Specifications
| PLA2G12A | |
| Polyclonal | |
| Unconjugated | |
| PLA2G12A | |
| 2310004B05Rik; FKSG38; group XII secreted phospholipase A2; group XII-1 phospholipase A2; group XIIA secreted phospholipase A2; group XIIA secretory phospholipase A2; GXII; GXII sPLA2; GXII-1; GXII-1-PLA2; mGXII-1; phosphatidylcholine 2-acylhydrolase 12A; phospholipase A2 group XIIA; phospholipase A2, group XII; phospholipase A2, group XIIA; Pla2g12; PLA2G12A; Rossy; sPLA2-XII; UNQ2519/PRO6012 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 362039 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| PLA2G12A | |
| synthetic peptide directed towards the middle region of rat PLA2G12A. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction